Order Entry
United States
ContactUsLinkComponent
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml), AAT Bioquest
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml), AAT Bioquest
Catalog # 76485-290
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml), AAT Bioquest
Catalog # 76485-290
Supplier:  AAT Bioquest
Supplier Number:  20005
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    FluoroQuest™ Anti-fading Mounting Medium with DAPI
  • Size:
    20 ml
  • Cat. No.:
    76485-290

Specifications

About this item

FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Preserves fluorescence
  • Prevents photobleaching
  • Contains nuclear counterstain for convenience
Caution: For research use only.