ContactUsLinkComponent
StrandBrite™ Green Fluorimetric RNA Quantitation Kit, AAT Bioquest
StrandBrite™ Green Fluorimetric RNA Quantitation Kit, AAT Bioquest
  76482-950
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
StrandBrite™ Green Fluorimetric RNA Quantitation Kit, AAT Bioquest
  76482-950
 :  17656
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    StrandBrite™ green fluorimetric RNA quantitation kit
  • Size:
    100 tests
  • Cat. No.:
    76482-950
  • No. of tests:
    100
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
 : For research use only.