- Description:StrandBrite™ green fluorimetric RNA quantitation kit *optimized for microplate readers*
- Size:1000 tests
- Cat. No.:76482-948
- No. of tests:1000
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
- Highly sensitive RNA detection
- Robust protocol
- Multiple applications