ContactUsLinkComponent
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole), AAT Bioquest
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole), AAT Bioquest
Catalog # 76485-678
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole), AAT Bioquest
Catalog # 76485-678
Supplier:  AAT Bioquest
Supplier Number:  17207
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    ddCTP [2',3'-dideoxycytidine-5'-triphosphate] *10 mm in ddH₂O*
  • Size:
    1 µmole
  • Cat. No.:
    76485-678
Specifications
About this item
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.