
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
Catalog # 76485-216
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:1 mg
- Cat. No.:76485-216
Specifications
About this item
:
:808.69 g/mol
ADR:5,II
:GHS07
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969