Rat Recombinant CD23/FCER2/FCER2A Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-482
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:Q6AZ45
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CLEC4J|BLAST-2|CLEC6|CLEC-6|CLECSF8|FCE2|CD23A|MCL|CD23|IGEBF|MPCL|MGC40078
- Protein/Peptide Name:FCER2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat FCER2 comprises 542 amino acids and predicts a molecular mass of 60.2 kDa. The apparent molecular mass of the recombinant protein is approximately 61 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Glu50-Pro331
- Shipping Temperature:Ambient
- Cat. No.:103666-482
Specifications
About this item
A DNA sequence encoding the rat FCER2 (Q6AZ45) (Glu50-Pro331) was expressed with Fc region of human IgG1 at the N-terminus.