Rat Recombinant E-Selectin/CD62e/SELE Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-268
Supplier: Sino Biological
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells. When 5 x 10E4 cells/well are added to rat E Selectin/Fc Chimera coated plates (2 μg/ml, 100 μl/well), approximately 30%-60% will adhere after 1 hour at 37 ℃. Optimal dilutions should be determined by each laboratory for each application.
- Gene ID:P98105-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ELAM|ELAM1|ESEL|LECAM2|CD62E
- Protein/Peptide Name:SELE
- Purity:>97% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat SELE comprises 484 amino acids and predicts a molecular mass of 53 kDa. The apparent molecular mass of the rat SELE is approximately 70-80 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Pro494
- Shipping Temperature:Ambient
- Cat. No.:103666-268
Specifications
About this item
A DNA sequence encoding the rat SELE (P98105-1) extracellular domain (Met 1-Pro 494) was expressed, fused with a polyhistidine tag at the C-terminus.