Order Entry
United States
ContactUsLinkComponent
Canine Recombinant TrkA/NTRK1 Protein (His Tag) (from HEK293 Cells)
Canine Recombinant TrkA/NTRK1 Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-156
Supplier:  Sino Biological
CAS Number:  
undefined
Canine Recombinant TrkA/NTRK1 Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-156
Supplier:  Sino Biological
Supplier Number:  70101-D07H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Dog
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to inhibit NGF-induced proliferation of TF‑1 human erythroleukemic cells.
    The ED50 for this effect is typically 3-24 µg/ml in the presence of 10 ng/ml of recombinant human NGF.
  • Gene ID:
    XP_851619.3
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TRKA|Trk-A|MTC|TRK1|TRK|p140-TrkA
  • Protein/Peptide Name:
    NTRK1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant canine NTRK1 comprises 146 amino acids and has a predicted molecular mass of 16.2 kDa. The apparent molecular mass of the protein is approximately 27-33 kDa in SDS-PAGE under reducing conditions due to glycosylation.
  • Sequence:
    Pro285-Lys410
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103666-156

Specifications

About this item

A DNA sequence encoding the canine NTRK1 (XP_851619.3) (Pro285-Lys410) was expressed with a N-terminal polyhistidine tag.