Aequorea victoria Recombinant Green fluorescent/enhanced GFP GFPSparkTM Protein (His Tag) (from E. coli)
Catalog # 103668-840
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Jellyfish
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:AAB65663
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:GFP
- Protein/Peptide Name:GFP
- Purity:>97% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Aequorea victoria GFPSparkTM consisting of 245 amino acids and has a calculated molecular mass of 27.7 kDa.
- Sequence:Met1-Leu238
- Shipping Temperature:Ambient
- Cat. No.:103668-840
Specifications
About this item
A DNA sequence encoding the Aequorea victoria GFP (AAB65663) (Met1-Leu238, except for the point mutations of intellectual property :F64L, S65T, R80K, S99F, T153M, I167V, H231L) was expressed, with a polyhistide tag at the C-terminus (Patent 201410599023.9).