Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant EphA3 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant EphA3 Protein (His Tag) (from HEK293 Cells)
Catalog # 103665-796
Supplier:  Sino Biological
CAS Number:  
undefined
Mouse Recombinant EphA3 Protein (His Tag) (from HEK293 Cells)
Catalog # 103665-796
Supplier:  Sino Biological
Supplier Number:  51122-M08H-200
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    200 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its binding ability in a functional ELISA.
    2. Immobilized mouse EPHA3-His at 10 μg/ml (100 μl/well) can bind mouse EFNA5-Fc (Cat:50597-M02H). The EC50 of mouse EFNA5-Fc (Cat:50597-M02H) is 4.9-11.4 ng/ml.
  • Gene ID:
    NP_034270.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Mek4|ETK1|Tyro4|Cek4|End3|Hek|Hek4|AW492086
  • Protein/Peptide Name:
    EphA3
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse EPHA3 comprises 532 amino acids and has a predicted molecular mass of 60.2 kDa.
  • Sequence:
    Met1-His541
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103665-796

Specifications

About this item

A DNA sequence encoding the mouse EPHA3 (NP_034270.1) (Met1-His541) was expressed with a C-terminal polyhistidine tag.