Mouse Recombinant FLT3L/Flt3 ligand Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse FLT3L (P49772-1) (Met1-Arg188) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured in a cell proliferation assay using BaF3 mouse proB cells transfected with mouse Flt3. The ED50 for this effect is typically 7-30 ng/ml.
- Gene ID:P49772-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:FLT3L|Ly72L|Flt3lg
- Protein/Peptide Name:FLT3LG
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse FLT3L /Fc is a disulfide-linked homodimer. The reduced monomer comprises 403 amino acids and has a predicted molecular mass of 45.4 kDa. The apparent molecular mass of the protein is approximately 56 and 35 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Arg188
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together