Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Tie2/CD202b/TEK Protein (ECD, Fc Tag) (from HEK293 Cells)
Mouse Recombinant Tie2/CD202b/TEK Protein (ECD, Fc Tag) (from HEK293 Cells)
Catalog # 103665-716
Supplier:  Sino Biological
CAS Number:  
undefined
Mouse Recombinant Tie2/CD202b/TEK Protein (ECD, Fc Tag) (from HEK293 Cells)
Catalog # 103665-716
Supplier:  Sino Biological
Supplier Number:  51087-M02H-200
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    200 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    1. Measured by its binding ability in a functional ELISA.
    2. Immobilized mouse ANGPT2 (Cat:50298-M07H) at 10 μg/ml (100 μl/well) can bind mouse TEK-Fc. The EC50 of mouse TEK-Fc is 0.58-1.34 μg/ml.
  • Gene ID:
    NP_038718.2
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TIE-2|VMCM|TIE2|CD202B|VMCM1
  • Protein/Peptide Name:
    TEK
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse Tek consists 964 amino acids and predicts a molecular mass of 107.7 kDa.
  • Sequence:
    Met1-Lys744
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103665-716

Specifications

About this item

A DNA sequence encoding the mouse Tek (NP_038718.2) (Met1-Lys744) was expressed with the Fc region of human IgG1 at the C-terminus.