Mouse Recombinant R-Spondin 1/RSPO1 Protein (His Tag) (from CHO cells)
A DNA sequence encoding the full length of mouse RSPO1 (NP_619624.2) (Met 1-Gln 265) was fused with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to induce activation of β-catenin response in a Topflash Luciferase assay using HEK293T human embryonic kidney cells. The ED50 for this effect is typically 0.1 - 0.9 μg/ml in the presence of 5 ng/ml recombinant mouse Wnt3a.
- Gene ID:NP_619624.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:N
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Rspondin|R-spondin
- Protein/Peptide Name:RSPO1
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant mouse RSPO1 comprises 256 amino acids with a predicted molecular mass of 28.5 kDa. As a result of glycosylation, the apparent molecular mass of the protein is approximately 44 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gln265
- Shipping Temperature:Ambient
Frequently Bought Together