Influenza B (B/Hong Kong/05/1972) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Catalog # 103633-362
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Influenza B
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:ABF21280.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant HA1 subunit of the Influenza B virus (B/Hong Kong/05/1972) hemagglutinin consists of 355 amino acids and predicts a molecular mass of 38.6 kDa.
- Sequence:Met1-Arg359
- Shipping Temperature:Ambient
- Cat. No.:103633-362
Specifications
About this item
A DNA sequence encoding the Influenza B virus (B/Hong Kong/05/1972)hemagglutinin (ABF21280.1) (Met1-Arg359), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.