Influenza A H5N1 (A/chicken/Yamaguchi/7/2004) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Catalog # 103636-856
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H5N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:BAD89305.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The secreted recombinant HA subunit of influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) comprises 525 amino acids and has a predicted molecular mass of 60 kDa.
- Sequence:Met1-Glu340
- Shipping Temperature:Ambient
- Cat. No.:103636-856
Specifications
About this item
A DNA sequence encoding the influenza A virus (A/chicken/Yamaguchi/7/2004(H5N1)) hemagglutinin (BAD89305.1) (Met1-Glu340), termed as HA1, was expressed with a C-terminal polyhistidine tag.