Order Entry
United States
ContactUsLinkComponent
Human Recombinant APOM Protein (Fc Tag) (from HEK293 Cells)
Human Recombinant APOM Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant APOM Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    O95445
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    G3a|HSPC336|DADB-127H9.5|NG20|apo-M
  • Protein/Peptide Name:
    APOM
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human APOM/Fc is a disulfide-linked homodimeric protein. The reduced monomer consists of 407 amino acids and has a predicted molecular mass of 45.6 kDa. The apparent molecular mass of the reduced monomer is approximately 50 kDa in SDS-PAGE under reducing conditions due to glycosylation.
  • Sequence:
    Met1-Asn188
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together