Human Recombinant SUV420H2 Protein (His & GST Tag) (from E. coli)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the amino acid sequence (Gly 2-Leu 280) of human SUV420H2 (NP_116090.2) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information
- Gene ID:NP_116090.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:KMT5C
- Protein/Peptide Name:KMT5C
- Purity:>80% as determined by SDS-PAGE
- Molecular Weight:The recombinant human SUV420H2/GST chimera consists of 516 amino acids and has a calculated molecular mass of 60 kDa as estimated in SDS-PAGE under reducing conditions.
- Sequence:Gly2-Leu280
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together