Human Recombinant LIFR/CD118 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the extracellular domain of human LIFR (NP_001121143.1) (Met 1-Ser 833) with a C-terminal polyhistidine tag was expressed.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:−20…−80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized LIF Protein, Human, Recombinant (hFc Tag)(Cat:14890-H02H) at 2 μg/ml (100 μl/well) can bind LIFR Protein, Human, Recombinant (His Tag)(Cat:10628-H08H), the EC50 is 60 - 360 ng/ml
- Gene ID:NP_001121143.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:N
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:SWS|SJS2|CD118|STWS|LIF-R
- Protein/Peptide Name:LIFR
- Purity:≥90% as determined by SDS-PAGE, ≥90% as determined by SEC-HPLC
- Molecular Weight:The secreted recombinant human LIFR comprises 800 amino acids with a predicted molecular mass of 91 kDa. As a result of glycosylation, rhLIFR migrates as an approximately 125-135 kDa band in SDS-PAGE under reducing conditions
- Sequence:Met1-Ser833
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together