Mouse Recombinant MERTK/Mer (His & GST Tag) Protein (from Baculovirus Insect Cells)
103665-106
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:The specific activity was determined to be 15 nmol/min/mg using Poly(Glu,Tyr) 4:1 as substrate
- Gene ID:Q60805
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:c-mer|RP38|Tyro12|MER|c-Eyk
- Protein/Peptide Name:MERTK
- Purity:>96% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse MERTK/GST chimera consists of 532 amino acids and has a calculated molecular mass of 61.7 kDa. The recombinant protein migrates as an approximately 58 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Glu573-Tyr867
- Cat. No.:103665-106
A DNA sequence encoding the mouse MERTK (Q60805) (Glu573-Tyr867) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
- Shipping temperature: Dry ice