Human Recombinant TNFSF14/LIGHT/CD258 Protein (Fc Tag) (from HEK293 Cells)
103632-600
:
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured in a cell proliferation assay using human umbilical vein endothelial cells (HUVEC). The ED50 for this effect is typically 5-25 ng/ml.
- Gene ID:AAK26160.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD258|LTg|TR2|LIGHT|HVEML
- Protein/Peptide Name:TNFSF14
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human TNFSF14 consists of 418 amino acids and predicts a molecular mass of 45.7 kDa.
- Sequence:Asp74-Val240
- Shipping Temperature:Ambient
- Cat. No.:103632-600
A DNA sequence encoding the human TNFSF14 (AAK26160.1) (Asp74-Val240) was expressed with the Fc region of rabbit IgG at the N-terminus..