Rhesus Recombinant HER2/ErbB2 Protein (His Tag) (from HEK293 Cells)
Catalog # 103585-086
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Rhesus Her2/ERBB2 His(Cat:90020-K08H) at 2 μg/ml (100 μl/well) can bind Herceptin, the EC50 of Herceptin is 10-50 ng/ml.
- Gene ID:XP_001090430.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:NEU|MLN19|HERVW|TKR1|HERV7Q|MLN 19|CD340|ERVWE1|HER2|HERV-W-ENV|ENV|HER-2/neu|HER-2|ENVW|HERVWENV|HERV-7q|NGL
- Protein/Peptide Name:ERBB2
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
- Molecular Weight:The recombinant Rhesus ErbB2 consists of 640 amino acids and predicts a molecular mass of 70.7 kDa. As a result of glycosylation, rhesus ErbB2 migrates as an approximately 105 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Thr652
- Shipping Temperature:Ambient
- Cat. No.:103585-086
Specifications
About this item
A DNA sequence encoding the Rhesus ErbB2 (XP_001090430.1) extracellular domain (Met 1-Thr 652) was fused with a polyhistidine tag at the C-terminus.