Rat Recombinant IL1RA Protein (His Tag) (from E. coli)
Catalog # 103887-172
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:NP_071530.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:IL1F3|IL-1ra|IL-1RN|DIRA|IRAP|IL-1ra3|ICIL-1RA|MVCD4|IL1RA
- Protein/Peptide Name:IL1RN
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat IL1RN comprises 164 amino acids and has a calculated molecular mass of 19.01 kDa.
- Sequence:His27-Gln178
- Shipping Temperature:Ambient
- Cat. No.:103887-172
Specifications
About this item
A DNA sequence encoding the rat IL1RN (NP_071530.1) (His27-Gln178) was expressed with a polyhistidine tag at the C-terminus.