Canine Recombinant FOLR1/Folate Receptor 1 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-196
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Dog
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:XP_851993.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:FOLR|Folate Binding Protein|FBP
- Protein/Peptide Name:FOLR1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant canine FOLR1 consists 442 amino acids and predicts a molecular mass of 50.7 kDa.
- Sequence:Met1-Met228
- Shipping Temperature:Ambient
- Cat. No.:103666-196
Specifications
About this item
A DNA sequence encoding the canine FOLR1 (XP_851993.1) (Met1-Met228) was expressed with the Fc region of human IgG1 at the C-terminus.