Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL27/Interleukin-27 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant IL27/Interleukin-27 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant IL27/Interleukin-27 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its binding ability in a functional ELISA. Immobilized mouse IL27-His at 10 μg/ml (100 μl/well) can bind human IL27RA-Fc (Cat:10489-H02H), The EC50 of human IL27RA-Fc (Cat:10489-H02H) is 0.26-0.62 μg/ml.
    2. Measured in antiviral assay using HepG2 cells infected with vesicular stomatitisvirus (VSV). The ED50 for this effect is 2-10ng/ml.
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    IL27p28|IL-27A|IL27A|IL-27|IL30|p28
  • Protein/Peptide Name:
    IL27
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant mouse IL27 comprises 442 amino acids and has a predicted molecular mass of 49.5 kDa. The apparent molecular mass of the protein is approximately 60 kDa in SDS-PAGE under reducing conditions due to glycosylation.
  • Sequence:
    Met1-Pro228 & Phe29-Ser234
  • Formulation:
    Lyophilized from sterile PBS, pH 7.4
  • Shipping Temperature:
    Ambient
  • Gene ID:
    O35228 & Q8K3I6

Specifications

Related Information

Frequently Bought Together