Mouse Recombinant MKK4/MEK4/MAP2K4 (His & GST Tag) Protein (from Baculovirus Insect Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse MAP2K4 (P47809) (Ala 2-Asp 397) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:No Kinase Activity
- Gene ID:P47809
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Serk1|MKK4|PRKMK4|Sek1|MEK4|JNKK1
- Protein/Peptide Name:MAP2K4
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The secreted recombinant mouse MAP2K4/GST chimera consists of 633 amino acids and has a calculated molecular mass of 72 kDa. The recombinant protein migrates as an approximately 65 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Ala2-Asp397
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together