Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Vitronectin/VTN Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant Vitronectin/VTN Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant Vitronectin/VTN Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by the ability of the immobilized protein to support the adhesion of DU145 human prostate carcinoma cells.
    When cells are added to mouse Vitronectin coated plates (10 μg/ml and 100 μl/well), > 60% cells will adhere specifically after 30 minutes at 37 ℃.
  • Gene ID:
    NP_035837.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Vitronectin|VNT|V75|VN
  • Protein/Peptide Name:
    VTN
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The secreted recombinant mouse vitronectin consists of 470 amino acids and has a predicted molecular mass of 54.2 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rmvitronectin is approximately 75-85 kDa due to glycosylation.
  • Sequence:
    Met1-Lys478
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together