Mouse Recombinant S100A4 Protein (Fc Tag) (from HEK293 Cells)
A DNA sequence encoding the mouse S100A4 (P07091) (Ala2-Lys101) was expressed with the Fc region of human IgG1 at the N-terminus.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P07091
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:pk9a|Capl|PeL98|FSp1|42a|metastasin|Mts1|18A2
- Protein/Peptide Name:S100A4
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse S100A4/Fc is a disulfide-linked homodimer. The reduced monomer comprises 360 amino acids and has a predicted molecular mass of 40 kDa. The apparent molecular mass of the protein is approximately 38-42 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Ala2-Lys101
- Shipping Temperature:Ambient
Frequently Bought Together