Mouse Recombinant CD62L/L-Selectin/SELL Protein (His & Fc Tag) (from HEK293 Cells)
Catalog # 103663-662
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells. When cells are added to SELL coated plates (10 μg/ml, 100 μl/well) approximately >60% cells will adhere specifically.
- Gene ID:NP_035476.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Protein/Peptide Name:SELL
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse SELL/Fc is a disulfide-linked homodimer after removal of the signal peptide. The reduced monomer consists of 542 amino acids and has a predicted molecular mass of 61 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rm SELL/Fc monomer is approximately 100-110 kDa due to glycosylation.
- Sequence:Met1-Asn332
- Shipping Temperature:Ambient
- Cat. No.:103663-662
Specifications
About this item
A DNA sequence encoding the extracellular domain (Met 1-Asn 332) of mouse SELL (NP_035476.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.