Order Entry
United States
ContactUsLinkComponent
Human Recombinant PTPRA/PTPalpha Protein (aa 174-793, His & GST Tag) (from Baculovirus Insect cells)
Human Recombinant PTPRA/PTPalpha Protein (aa 174-793, His & GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant PTPRA/PTPalpha Protein (aa 174-793, His & GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to bind biotinylated recombinant mouse SRC in a functional ELISA.
    2. Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP). The specific activity is >40000 pmol/min/μg.
  • Gene ID:
    P18433-2
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    PTPRL2|RPTPA|HPTPalpha|PTPA|HPTPA|R-PTP-alpha|HEPTP|HLPR|LRP
  • Protein/Peptide Name:
    PTPRA
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human PTPRA (aa 174-793)/GST chimera consists of 857 amino acids and has a calculated molecular mass of 99 kDa. It migrates as an approximately 90 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Ala174-Lys793
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together