Influenza A H5N1 (A/chicken/India/NIV33487/06) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Catalog # 103632-732
Supplier: Sino Biological
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H5N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABQ45850.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant HA1 subunit of influenza A H5N1 HA (A/chicken/India/NIV33487/2006 (H5N1)) comprises 335 amino acids and has a predicted molecular mass of 38 kDa. As a result of glycosylation, it migrates as an approximately 45-50 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Glu340
- Shipping Temperature:Ambient
- Cat. No.:103632-732
Specifications
About this item
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza A hemagglutinin (A/chicken/India/NIV33487/06 (H5N1)) (ABQ45850.1), termed as HA1, was fused with a C-terminal polyhistidine tag.