Human Recombinant SRC Kinase/Proto-oncogene c-Src Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human SRC (P12931-1) (Met 1-Leu 536) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:The specific activity was determined to be >80 nmol/min/mg using Poly(Glu:Tyr) 4:1 as substrate.
- Gene ID:P12931-1
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ASV|p60-Src|c-SRC|SRC1
- Protein/Peptide Name:SRC
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human SRC/GST chimera consists of 773 amino acids and has a calculated molecular mass of 87.7 KDa. It migrates as an approximately 81 KDa band in SDS-PAGE under non-reduced conditions.
- Sequence:Met1-Leu536
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together