Human Recombinant Complement Factor H/CFH Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the C-terminal segment of CFH isoform a (NP_000177.2), corresponding to amino acid (Ser 860-Arg 1231) was expressed, fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind biotinylated human DMP1 in a functional ELISA.
- Gene ID:NP_000177.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HUS|ARMS1|HF2|FH|HF|CFHL3|AMBP1|HF1|FHL1|ARMD4|AHUS1
- Protein/Peptide Name:CFH
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human CFH consists of 383 amino acids and has a calculated molecular mass of 43 kDa. The apparent molecular mass of rh CFH is approximately 55-60 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Ser860-Arg1231
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together