Order Entry
United States
ContactUsLinkComponent
 
Rat Recombinant ALK-1/ACVRL1 Protein (His & Fc Tag) (from HEK293 Cells)
  103621-342
 :  
undefined
Rat Recombinant ALK-1/ACVRL1 Protein (His & Fc Tag) (from HEK293 Cells)
  103621-342
 :  10596-R03H-200
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    200 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to inhibit BMP9-induced alkaline phosphatase production by MC3T3E1 mouse chondrogenic cells. David, L. et al. (2007) Blood 109:1953.
    The ED50 for this effect is typically 40-200 ng/ml in the presence of 2 ng/ml of recombinant human BMP9.
  • Gene ID:
    NP_071886.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    HHT2|SKR3|ALK-1|TSR-I|ALK1|ORW2|ACVRLK1|HHT
  • Protein/Peptide Name:
    ACVRL1
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The secreted recombinant rat ACVRL1/Fc is a disulfide-linked homodimer after removal of the signal peptide. The reduced monomer comprises 343 amino acids with a predicted molecular mass of 38.7 kDa. As a result of glycosylation, it migrates as an approximately 50-55 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ala118
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103621-342

 

 

A synthetic DNA sequence encoding the rat ACVRL1 (NP_071886.1) extracellular domain (Met 1-Ala 118) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.