Human Recombinant NCR3 (CD337) Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103906-252
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_001138938.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD337|DAAP-90L16.3|1C7|LY117|NCR3|NKp30|MALS
- Protein/Peptide Name:NCR3
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human NCR3 consists of 355 amino acids and predicts a molecular mass of 39.6 kDa.
- Sequence:Met1-Gly135
- Shipping Temperature:Ambient
- Cat. No.:103906-252
Specifications
About this item
A DNA sequence encoding the human NCR3 (NP_001138938.1) (Met1-Gly135) was expressed with the Fc region of human IgG1 at the C-terminus.