Human VTN / Vitronectin Protein (His Tag)
: Avantor
A DNA sequence encoding the human VTN (NP_000629.3) (Met1-Leu478) was expressed with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_000629.3
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Vitronectin|VNT|V75|VN
- Protein/Peptide Name:VTN
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:53.7 kDa
- Sequence:Met1-Leu478
- Shipping Temperature:Ambient
Frequently Bought Together