Human Recombinant IL12A/NKSF1 Protein (Fc Tag) (from HEK293 Cells)
A DNA sequence encoding the p35 subunit of human IL12, termed as IL12A (P29459) (Met 1-Ser 219) was fused with the Fc region of human IgG1 at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P29459
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CLMF|IL-12A|P35|NFSK|NKSF1
- Protein/Peptide Name:IL12A
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human IL12A/Fc is a disulfide-linked homodimer. The reduced monomer consists of 435 amino acids and has a predicted molecular mass of 49.3 kDa. As a result of glycosylation, the rhIL12A/Fc monomer migrates as an approximately 65-70 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ser219
- Shipping Temperature:Ambient
Frequently Bought Together