Mouse Recombinant TIGIT Protein (mFc Tag) (from HEK293 Cells)
103886-916
:
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:50 µg
- Environmentally Preferable:
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Immobilized Mouse CD155/PVR Protein, Recombinant (His Tag)(HPLC-50259-M08H) at 2 μg/ml (100 μl/well) can bind TIGIT Protein, Mouse, Recombinant (mFc Tag)(50939-M38H), the EC50 of TIGIT Protein, Mouse, Recombinant (mFc Tag)(50939-M38H) is 10-50 ng/ml.
- Gene ID:NP_001139797.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:WUCAM|VSTM3|VSIG9
- Protein/Peptide Name:TIGIT
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse TIGIT consists of 358 amino acids and predicts a molecular mass of 39.93 kDa.
- Sequence:Met1-Gly141
- Shipping Temperature:Ambient
- Cat. No.:103886-916
A DNA sequence encoding the mouse TIGIT (NP_001139797.1) (Met1-Gly141) was expressed with the Fc region of mouse IgG2a at the C-terminus.