Mouse Recombinant CD68 Protein (His Tag) (from Baculovirus Insect Cells)
A DNA sequence encoding the mouse CD68 (NP_001277987.1) (Met1-Ser291) was expressed with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_001277987.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:N
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:SCARD1|LAMP4|gp110
- Protein/Peptide Name:CD68
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse CD68 consists of 282 amino acids and predicts a molecular mass of 30.2 kDa.
- Sequence:Met1-Ser291
- Shipping Temperature:Ambient
Frequently Bought Together