Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Noggin/NOG Protein (Fc Tag) (from HEK293 Cells)
Mouse Recombinant Noggin/NOG Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant Noggin/NOG Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to inhibit BMP4-induced alkaline phosphatase production by MC3T3-E1 cells. The ED50 for this effect is typically 0.1-0.6 μg/ml.
  • Gene ID:
    P97466
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    SYM1|SYNS1|Noggin
  • Protein/Peptide Name:
    NOG
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse NOG/Fc comprises 454 amino acids and has a predicted molecular mass of 51 kDa. It migrates as approximately 60 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Cys232
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together