Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 (2019-nCoV) 3CLpro/3C-like protease-His & AVI Recombinant Protein (from E. coli)
SARS-CoV-2 (2019-nCoV) 3CLpro/3C-like protease-His & AVI Recombinant Protein (from E. coli)
Catalog # 103886-248
Supplier:  Sino Biological
CAS Number:  
SARS-CoV-2 (2019-nCoV) 3CLpro/3C-like protease-His & AVI Recombinant Protein (from E. coli)
Catalog # 103886-248
Supplier:  Sino Biological
Supplier Number:  40594-V56E-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    SARS-CoV-2
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    Please contact us for more information.
  • Biological Activity:
    Active
  • Gene ID:
    YP_009725295.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    3CLpro
  • Protein/Peptide Name:
    3CLpro
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant SARS-CoV-2 (2019-nCoV) 3CL Protease consists of 328 amino acids and predicts a molecular mass of 36.57 kDa.
  • Sequence:
    Ser3264-Gln3569
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103886-248

Specifications

About this item

A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) 3CL Protease (YP_009725295.1) (Ser3264-Gln3569) was expressed with a N-terminal polyhistidine tagged AVI tag at the N-terminus.