Influenza B (B/Yamagata/16/1988) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-852
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Influenza B
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured by its ability to agglutinate guinea pig red blood cells. HA titer is 1-10 μg/ml for 1%GRBC
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of Influenza B virus (B/Yamagata/16/1988) consists 542 amino acids and predicts a molecular mass of 59 kDa.
- Sequence:Met1-Thr546
- Shipping Temperature:Ambient
- Cat. No.:103663-852
Specifications
About this item
A DNA sequence encoding the Influenza B virus (B/Yamagata/16/1988) hemagglutinin (Met1-Thr546), termed as HA, was expressed with a polyhistidine tag at the C-terminus.