A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32a (AAL92096.1) extracellular domain (Met 1-Gly 220) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized Cynomolgus CD32A Protein (His & AVI Tag), Biotinylated (Cat:90015-C27H-B) at 2 μg/ml (100 μl/well) can bind Human IgG2 Protein (257 Ser/Ala) (Cat:13504-HNAH), the EC50 is 15-140 ng/ml.
- Gene ID:AAL92096.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD32|FcGR|CD32A|IGFR2|Fc gamma RIIA|FCG2|FCGR2A1|CDw32|FCGR2
- Protein/Peptide Name:FCGR2A
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant cynomolgus CD32a consists of 206 amino acids and has a calculated molecular mass of 22.9 kDa. The apparent molecular mass of the recombinant protein is approximately 33-35 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Ala220
- Shipping Temperature:Ambient