Rat Recombinant ICOS Ligand/B7-H2 Protein (His Tag) (from HEK293 Cells)
Catalog # 103670-066
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:XP_006223893.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ICOS-L|ICOS ligand|B7RP-1|ICOSLG|CD275|LICOS|B7-h2|B7RP1|ICOSL|GL50|B7H2
- Protein/Peptide Name:ICOSLG
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat ICOSLG consists 250 amino acids and predicts a molecular mass of 28.6 kDa.
- Sequence:Met1-Lys261
- Shipping Temperature:Ambient
- Cat. No.:103670-066
Specifications
About this item
A DNA sequence encoding the rat ICOSLG (XP_006223893.1) (Met1-Lys261) was expressed with a polyhistidine tag at the C-terminus.