Rat Recombinant IL1R2/IL1RB/CD121b Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-450
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P43303
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:IL-1RT2|IL1RB|CD121b|CDw121b|IL-1R-2|IL1R2c|IL-1RT-2
- Protein/Peptide Name:IL1R2
- Purity:>97% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat IL1R2 comprises 353 amino acids and predicts a molecular mass of 39.7 kDa. The apparent molecular mass of the rat IL1R2 is approximately 55-60 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Glu355
- Shipping Temperature:Ambient
- Cat. No.:103666-450
Specifications
About this item
A DNA sequence encoding the rat IL1R2 (P43303) extracellular domain (Met 1-Glu 355) was expressed, fused with a polyhistidine tag at the C-terminus.