Order Entry
United States
ContactUsLinkComponent
 
Rat Recombinant TNFR1/CD120a/TNFRSF1A Protein (Fc Tag) (from HEK293 Cells)
  103666-414
 :  
undefined
Rat Recombinant TNFR1/CD120a/TNFRSF1A Protein (Fc Tag) (from HEK293 Cells)
  103666-414
 :  80181-R02H-100
 :  

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to inhibit TNFα-mediated cytotoxicity in L-929 mouse fibroblast cells in the presence of metabolic inhibitor actinomycin D. The ED50 for this effect is typically 3-30 ng/ml in the presence of 0.05 ng/ml of ratTNFα.
    2. Immobilized mouse TNFa (80-235) (Cat:50349-MNAE ) at 10 μg/ml (100 μl/well) can bind rat TNFRSF1A-Fc, The EC50 of rat TNFRSF1A-Fc is 0.08-0.20 μg/ml.
  • Gene ID:
    P22934
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TNFR1-d2|p55-R|TNF-R|TNF-R-I|TBP1|TNFR55|p55|CD120a|MS5|FPF|TNFAR|p60|TNFR1|TNFR60|TNF-R55
  • Protein/Peptide Name:
    TNFRSF1A
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant rat TNFRSF1A/Fc is a disulfide-linked homodimer. The reduced monomer comprises 431 amino acids and has a predicted molecular mass of 47.9 kDa. The apparent molecular mass of the protein is approximately 59 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ala211
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103666-414

 

 

A DNA sequence encoding the rat TNFRSF1A (P22934) (Met1-Ala211) was expressed, fused with the Fc region of human IgG1 at the C-terminus.