Rat Recombinant EphA4 Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-362
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:200 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its binding ability in a functional ELISA.
Immobilized recombinant Rat EPHA4 Protein (His Tag) (Cat: 80123-R08H) at 10 µg/ml (100 μl/well) can bind Recombinant rat EFNA5-Fc (Cat: 80105-R02H) with a linear range of 7.8-12.5 ng/ml. - Gene ID:D3ZZK3
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Sek1|Cek8|AI385584|Tyro1|rb|2900005C20Rik|Hek8|Sek
- Protein/Peptide Name:EphA4
- Purity:>97% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat EPHA4 comprises 539 amino acids and predicts a molecular mass of 59.8 kDa. The apparent molecular mass of the recombinant protein is approximately 71 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Thr547
- Shipping Temperature:Ambient
- Cat. No.:103666-362
Specifications
About this item
A DNA sequence encoding the rat EPHA4 (D3ZZK3) (Met1-Thr547) was expressed, fused with a polyhistidine tag at the C-terminus.