Rat Recombinant GFRA1/GFR alpha-1 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-234
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized rat His-GDNF (78-211) (Cat:80050-R07B) at 10 μg/ml (100 μl/well) can bind rat GFRA1-Fc, The EC50 of rat GFRA1-Fc is 20-46.6 ng/ml.
- Gene ID:Q62997
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:GFR-ALPHA-1|RETL1|GDNFR|RET1L|GDNFRA|TRNR1
- Protein/Peptide Name:GFRA1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat GFRA1/Fc is a disulfide-linked homodimer. The reduced monomer comprises 662 amino acids and has a predicted molecular mass of 73.8 kDa. The apparent molecular mass of the protein is approximately 85 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Leu445
- Shipping Temperature:Ambient
- Cat. No.:103666-234
Specifications
About this item
A DNA sequence encoding the rat GFRA1 (Q62997) (Met1-Leu445) was expressed with the Fc region of human IgG1 at the C-terminus.