Order Entry
United States
ContactUsLinkComponent
Canine Recombinant ALK-3/BMPR1A Protein (Fc Tag) (from HEK293 Cells)
Canine Recombinant ALK-3/BMPR1A Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103665-746
Supplier:  Sino Biological
CAS Number:  
undefined
Canine Recombinant ALK-3/BMPR1A Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103665-746
Supplier:  Sino Biological
Supplier Number:  70051-D02H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Dog
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    Measured by its ability to inhibit rhBMP4-induced alkaline phosphatase production by MC3T3-E1 cells.
    The ED50 for this effect is typically 0.5-2.5 μg/ml in the presence of 50 ng/ml of recombinant human BMP4.
  • Gene ID:
    NP_001138622.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    SKR5|ALK3|CD292|ACVRLK3
  • Protein/Peptide Name:
    BMPR1A
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant canine BMPR1A consists 370 amino acids and predicts a molecular mass of 41.2 kDa.
  • Sequence:
    Met1-Arg152
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103665-746

Specifications

About this item

A DNA sequence encoding the canine BMPR1A (NP_001138622.1) (Met1-Arg152) was expressed with the Fc region of human IgG1 at the C-terminus.