Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 (2019-nCoV) Recombinant NSP3-His Protein (from E. coli)
SARS-CoV-2 (2019-nCoV) Recombinant NSP3-His Protein (from E. coli)
Catalog # 103885-986
Supplier:  Sino Biological
CAS Number:  
SARS-CoV-2 (2019-nCoV) Recombinant NSP3-His Protein (from E. coli)
Catalog # 103885-986
Supplier:  Sino Biological
Supplier Number:  40638-V07E-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    SARS-CoV-2
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    YP_009725299.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    NSP3-CoV
  • Protein/Peptide Name:
    NSP3-CoV
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant SARS-CoV-2 (2019-nCoV) NSP3 comprises 182 amino acids and has a calculated molecular mass of 19.87 kDa.
  • Sequence:
    Ile205-Lys379
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103885-986

Specifications

About this item

A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) NSP3 (YP_009725299.1) (Ile205-Lys379) was expressed with a polyhistidine tag at the N-terminus.