Order Entry
United States
ContactUsLinkComponent
Influenza A H5N1 (A/barnswallow/HongKong/D10-1161/2010) Recombinant Hemagglutinin (HA1+HA2) Protein (His Tag) (from Baculovirus Insect Cells)
Influenza A H5N1 (A/barnswallow/HongKong/D10-1161/2010) Recombinant Hemagglutinin (HA1+HA2) Protein (His Tag) (from Baculovirus Insect Cells)
  103633-056
 :  
undefined
Influenza A H5N1 (A/barnswallow/HongKong/D10-1161/2010) Recombinant Hemagglutinin (HA1+HA2) Protein (His Tag) (from Baculovirus Insect Cells)
  103633-056
 :  40160-V08B1-100
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    H5N1
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to bind with Neu5Aca2-3Galb1-4GlcNAcb-PAA-biotin (01-077) using the Octet RED System.
    2. Measured by its ability to agglutinate guinea pig red blood cells. HA titer is 2-8 ng/ml for 1% GRBC.
  • Gene ID:
    AGC13463.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hemagglutinin
  • Protein/Peptide Name:
    HA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant hemagglutinin of Influenza A virus (A/barnswallow/HongKong/D10-1161/2010(H5N1)) comprises 524 amino acids and has a predicted molecular mass of 59.8 kDa. The apparent molecular mass of the protein is approximately 58.3 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Gln531
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103633-056

 

 

A DNA sequence encoding the Influenza A virus (A/barnswallow/HongKong/D10-1161/2010(H5N1)) hemagglutinin (AGC13463.1) (Met1-Gln531) with cleavage site mutated (RRRRK →TET, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.